Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-USP1/His6-UAF1 Complex Protein

Catalog No. E568050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E568050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E568050 Supplier R&D Systems™ Supplier No. E568050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Recombinant Human USP1/UAF1 complex is a Ubiquitin-specific deconjugating enzyme heterodimer. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human USP1/UAF1 complex concentration of 0.01-0.5 μM.

Specifications

Accession Number O94782.1 (USP1), Q8TAF3.1 (UAF1)
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, TCEP and Glycerol.
Gene ID (Entrez) 7398
Molecular Weight (g/mol) M.W. (theoretical): 89 kDa (USP1), 77 kDa (UAF1)
Name His6 USP1/UAF1 Complex
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP1/UAF1 Complex protein Met1 - Leu785 (Gly670Ala, Gly671Ala) with a C-terminal 6-His tag (USP1), Met1 - Thr677 with a C-terminal 6-His tag (UAF1)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol USP1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein USP1/UAF1 Complex
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.