Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-USP1 Protein

Catalog No. E564050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E564050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E564050 Supplier R&D Systems™ Supplier No. E564050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Recombinant Human USP1 is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human USP1 concentration of 0.01-0.5 μM.

Specifications

Accession Number O94782
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES, pH 7.5, 100 mM NaCl, 10% glycerol, 1 mM TCEP
Gene ID (Entrez) 7398
Molecular Weight (g/mol) M.W. (theoretical): 89 kDa
Name His6 USP1
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP1 protein Contains substitutions at G670A & G671A and a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol USP1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein USP1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.