Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-USP15 Protein

Catalog No. E594050
Change view
Click to view available options
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
E594050 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E594050 Supplier R&D Systems™ Supplier No. E594050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human USP15 concentration of 5-20 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555).

Specifications

Accession Number Q9Y4E8
For Use With (Application) Bioactivity
Formulation X mg/mL (XμM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 1 mM TCEP, 10% (v/v) Glycerol
Gene ID (Entrez) 9958
Molecular Weight (g/mol) M.W. (theoretical): 113 kDa
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP15 protein Contains an C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias Deubiquitinating enzyme 15, EC 3.1.2.15, EC 3.4.19.12, Gcap18, KIAA0529, KIAA0529ubiquitin carboxyl-terminal hydrolase 15, MGC131982, MGC149838, MGC74854, ubiquitin specific peptidase 15, ubiquitin specific protease 15, ubiquitin thioesterase 15, Ubiquitin thiolesterase 15, Ubiquitin-specific-processing protease 15, Ubp109, unph-2, UNPH4
Common Name USP15
Gene Symbol USP15
Biological Activity Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human USP15 concentration of 5-20 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555).
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Contains an C-terminal 6-His tag
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein USP15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.