Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-USP30 Protein

Catalog No. E582050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E582050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E582050 Supplier R&D Systems™ Supplier No. E582050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Reaction conditions will need to be optimized for each specific application. We recommend an initial USP30 concentration of 20-100 nM in reactions utilizing Ubiquitin-AMC or Ubiquitin-Rhodamine substrates (U-550 or U-555), or 200-500 nM when digesting recombinant Polyubiquitin chain substrates.

Specifications

Accession Number Q70CQ3.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, Glycerol and DTT.
Gene ID (Entrez) 84749
Molecular Weight (g/mol) M.W. (theoretical): 54 kDa
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP30 protein Thr57 - Glu517 with a N-terminal Met and a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias Deubiquitinating enzyme 30, EC 3.1.2.15, EC 3.4.19.12, FLJ40511, MGC10702, ubiquitin carboxyl-terminal hydrolase 30, ubiquitin specific peptidase 30, ubiquitin specific protease 30, ubiquitin thioesterase 30, Ubiquitin thiolesterase 30, ubiquitin-specific protease 30, Ubiquitin-specific-processing protease 30, Ub-specific protease 30
Common Name USP30
Gene Symbol USP30
Biological Activity Reaction conditions will need to be optimized for each specific application. We recommend an initial USP30 concentration of 20-100 nM in reactions utilizing Ubiquitin-AMC or Ubiquitin-Rhodamine substrates (U-550 or U-555), or 200-500 nM when digesting recombinant Polyubiquitin chain substrates.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Contains a C-terminal 6-His tag, Thr57 - Glu517 Accession # Q70CQ3
Purity or Quality Grade >98%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein USP30
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.