Learn More
R&D Systems™ Recombinant Human HO-1/HMOX1 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl and Glycerol. |
| Gene ID (Entrez) | 3162 |
| Molecular Weight (g/mol) | 31 kDa |
| Name | HO-1/HMOX1/HSP32 |
| Quantity | 20 μg |
| Source | E. coli-derived human HO-1/HMOX1/HSP32 protein Met1-Thr261 (Phe33Leu),with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | AA Additional Info: Met1-Thr261 (Phe33Leu), with a C-terminal 6-His tag |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.