Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HSP70/HSPA1A His Protein
SDP

Catalog No. NBC118367 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118367 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118367 Supplier R&D Systems™ Supplier No. NBC118367

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-641 of Human HSP70/HSPA1A The Recombinant Human HSP70/HSPA1A Protein is derived from E. coli. The Recombinant Human HSP70/HSPA1A Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P08107
For Use With (Application) SDS-PAGE, Western Blot
Formulation 20mM Tris buffer (pH 7.5), 2mM DTT
Gene ID (Entrez) 3303
Molecular Weight (g/mol) 72.2kDa
Name HSP70/HSPA1A Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen HSP70 (amino acids 1-641) Human, Recombinant, His- tagged, E.coli (NP_005336).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.