Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HSP90 alpha Protein

Catalog No. AP160050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP160050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AP160050 Supplier R&D Systems™ Supplier No. AP160050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Reaction conditions will need to be optimized for each specific application. IMPORTANT: HSP90 alpha/HSP90AA1 works in conjunction with the co-chaperones such as p23/PTGES3--both proteins are typically required for enzymatic activity. For in vitro use we recommend an initial co-chaperone concentration of 2-3 µM, with an equimolar (or greater) concentration of HSP90 alpha/HSP90AA1.

Specifications

Accession Number P07900.5
Formulation Supplied as a 0.2 μm filtered solution in HEPES, KCl and TCEP.
Gene ID (Entrez) 3320
Molecular Weight (g/mol) 85 kDa
Name HSP90 alpha
Quantity 50 μg
Source E. coli-derived human HSP90 alpha protein Met1 - Asp732
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.