Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HSPA8/HSC71/Hsc70 His Protein
SDP

Catalog No. NBP130278 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130278 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130278 Supplier Novus Biologicals™ Supplier No. NBP130278
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-646 of Human HSPA8/HSC71/Hsc70 The Recombinant Human HSPA8/HSC71/Hsc70 Protein is derived from E. coli. The Recombinant Human HSPA8/HSC71/Hsc70 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P11142
Concentration 1 mg/mL
For Use With (Application) In vitro Assay, SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 10% glycerol
Gene ID (Entrez) 3312
Molecular Weight (g/mol) M.W. (theoretical): 73.1 kDa
Name HSPA8/HSC71/Hsc70 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Recombinant HSC70 protein was expressed in E.coli and purifiied (NP_006588).
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol HSPA8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Membrane Trafficking and Chaperones
Purity or Quality Grade >90%, by SDS-PAGE
Protein HSPA8/HSC71/Hsc70
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.