Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HTRA2/Omi Protein
SDP

Catalog No. 1458HT100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1458HT100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1458HT100 Supplier R&D Systems™ Supplier No. 1458HT100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave beta-casein. >95% cleavage of beta-casein, as measured under the described conditions.

Specifications

Accession Number O43464
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 27429
Molecular Weight (g/mol) M.W. (Observed): 38 kDa, reducing conditions; M.W. (theoretical): 36 kDa
Name HTRA2/Omi
Quantity 100 μg
Source E. coli-derived human HTRA2/Omi protein Ala134-Glu458, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Gene Symbol HTRA2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein HTRA2/Omi
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.