Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ICAM-1/CD54 Fc Chimera Avi-tag Protein

Catalog No. AVI720050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AVI720050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AVI720050 Supplier R&D Systems™ Supplier No. AVI720050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Biotinylated

  • Measured by its binding ability in a functional ELISA.
  • When Human ICAM-1 (14C11) Antibody is immobilized at 0.5 μg/mL (100 μL/well), Biotinylated Recombinant Human ICAM-1/CD54 Fc Chimera Avi-tag binds with an ED50 of 3 to 18 ng/mL.
  • Measured by its ability to support the adhesion of HSB2 human peripheral blood acute lymphoblastic leukemia cells.
  • The ED50 for this effect is 0.3 to 2.7 μg/mL.

Specifications

Accession Number P05362
For Use With (Application) Bioactivity
Gene ID (Entrez) 3383
Molecular Weight (g/mol) 78 kDa
Name BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human ICAM-1/CD54 protein

Human ICAM-1 (Gln28-Glu480) Accession # P05362IEGRMDHuman IgG1 (Pro100-Lys330)GGAvi-tag

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Biotin
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.