Learn More
R&D Systems™ Recombinant Human ICAM-1/CD54 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Trehalose. |
| Gene ID (Entrez) | 3383 |
| Molecular Weight (g/mol) | 50 kDa |
| Name | ICAM-1/CD54 |
| Quantity | 200 μg |
| Source | Mouse myeloma cell line,NS0-derived human ICAM-1/CD54 protein Met1-Glu480 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.