Learn More
R&D Systems™ Recombinant Human ICAM-4 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q14773.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3386 |
| Molecular Weight (g/mol) | M.W.-observed: 50-60 kDa & 70-80 kDa, under reducing conditions, M.W.-theoretical: 50 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human ICAM-4 protein Human ICAM-4 (Ala31-Ala240) (N-terminus) Accession # Q14773.1 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD242, CD242 antigen, ICAM4, ICAM-4, intercellular adhesion molecule 4, intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), intercellular adhesion molecule 4 (LW blood group), intercellular adhesion molecule 4, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group antigen a, Landsteiner-Wiener blood group glycoprotein, LWLW blood group protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.