Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ICOS (aa 21-140) mFc Chimera Protein, CF

Catalog No. 11423CS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-423-CS050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-423-CS050 Supplier R&D Systems™ Supplier No. 11423CS050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. Recombinant Human ICOS mFc Chimera (Catalog # 11423-CS) binds to Recombinant Human B7-H2 Fc Chimera Protein (Catalog # 165-B7 ) with a ED50 of 1.50-15.0 ng/mL.

Specifications

Accession Number Q9Y6W8.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 29851
Molecular Weight (g/mol) M.W. (Observed): 54-60 kDa, under reducing conditions., M.W. (Theroretical): 41 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human ICOS protein Human ICOS (Glu21-Lys140) (N-terminus) Accession # Q9Y6W8.1 IEGRMD Mouse IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ICOS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.