Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ICOS Fc Chimera Protein

Catalog No. 169CS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
169CS050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 169CS050 Supplier R&D Systems™ Supplier No. 169CS050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit human T cell proliferation induced by rhB7-H2/Fc Chimera in the presence of anti-CD3. Wang, S. et al. (2000) Blood 96:2808. The ED50 for this effect is 1-4 μg/mL in the presence of 3 μg/mL Recombinant Human B7-H2 Fc Chimera (Catalog # 165-B7 ) and at least 20 ng/mL of anti-CD3. Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number Q9Y6W8
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 29851
Molecular Weight (g/mol) M.W. (Observed): 50-60 kDa, reducing conditions; M.W. (theoretical): 40.3 kDa (monomer)
Name ICOS Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human ICOS protein Human ICOS (Glu21-Phe141) (N-terminus) Accession # Q9Y6W8 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol ICOS
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein ICOS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.