Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IDO2 Protein

Catalog No. 9967AO050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
99-67A-O050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 99-67A-O050 Supplier R&D Systems™ Supplier No. 9967AO050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Indoleamine 2,3-dioxygenase 2

Measured by its ability to oxidize L-tryptophan to N-formyl-kynurenine. The specific activity is >15 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q6ZQW0-1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 169355
Molecular Weight (g/mol) M.W. (Observed): 41 kDa, reducing conditions; M.W. (theoretical): 46 kDa
Quantity 50 μg
Source E. coli-derived human IDO2 protein Glu15-Gly420, with an N-terminal Met and C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity IDO2
Species Human
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IDO2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.