Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IFN-alpha 1 (Alpha D [Ala 114]) Protein
SDP

Catalog No. 111751 Shop All R&D Systems Products
Change view
Click to view available options
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
111751 Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 111751 Supplier R&D Systems™ Supplier No. 111751
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IFN-alpha 1 (Alpha D [Ala 114]) Protein is derived from E. coli. The Recombinant Human IFN-alpha 1 (Alpha D [Ala 114]) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number J00210
For Use With (Application) Bioactivity
Formulation Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA)
Molecular Weight (g/mol) Predicted MW: 19.4kDa
Source E. coli-derived human IFN-alpha 1 protein
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Conjugate Unconjugated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.