Learn More
Description
Specifications
Specifications
| Accession Number | P01562 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4, containing 3 % Mannitol, 0.05 % Tween-80. |
| Gene ID (Entrez) | 3439 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 19.5 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of IFN-alpha 1 AS determined by LAL method. |
| Gene Alias | 1, IFL;IFN;IFNA@;IFNA13;IFN-ALPHA;IFN-alphaD;Interferon alpha-1/13;leIF D |
| Gene Symbol | IFNA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
