Learn More
R&D Systems™ Recombinant Human IFN-alpha 2 (alpha 2b) Protein
Catalog No. 111051
This item is not returnable. View return policy
Catalog No. 111051 Supplier R&D Systems™ Supplier No. 111051
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene Alias alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765