Learn More
R&D Systems™ Recombinant Human IFN-alpha B2 (alpha 8) Protein
Catalog No. 111151
This item is not returnable. View return policy
Catalog No. 111151 Supplier R&D Systems™ Supplier No. 111151
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene Alias IFNA8, IFNalpha B2, IFN-alpha B2, IFN-alpha-8, IFN-alphaB, interferon alpha type 201, interferon alpha-8, interferon alpha-B', Interferon alpha-B, Interferon alpha-B2, interferon, alpha 8, LeIF B