Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IFN-alpha F (alpha 21) Protein

Catalog No. 111301R
Change view
Click to view available options
Quantity:
100,000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-130-1R 100,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-130-1R Supplier R&D Systems™ Supplier No. 111301
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

  • Alternate Names: IFNA21; IFN-alpha-21; IFN-alphaI; interferon alpha-21; Interferon alpha-F; interferon, alpha 21; LeIF F; leukocyte interferon protein; MGC126687; MGC126689

Specifications

Gene ID (Entrez) 3452 (Human)
Quantity 100,000 U
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Regulatory Status RUO
Gene Alias IFNA21, IFNalpha F, IFN-alpha F, IFN-alpha-21, IFN-alphaI, interferon alpha-21, Interferon alpha-F, interferon, alpha 21, LeIF F, leukocyte interferon protein, MGC126687, MGC126689
Conjugate Unconjugated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.