Learn More
R&D Systems™ Recombinant Human IFN-alpha G (alpha 5) Protein
Catalog No. 111351
This item is not returnable. View return policy
Catalog No. 111351 Supplier R&D Systems™ Supplier No. 111351
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.