Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IFN-beta Protein

Catalog No. 8499IF010CF
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-99I-F010CF 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-99I-F010CF Supplier R&D Systems™ Supplier No. 8499IF010/CF
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IFN-beta Protein is derived from CHO. The Recombinant Human IFN-beta Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P01574
For Use With (Application) Bioactivity
Gene ID (Entrez) 3456
Molecular Weight (g/mol) Observed M.W.: 20-26kDa, reducing conditions, Theoretical M.W.: 20kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human IFN-beta protein Met22-Asn187
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Common Name IFN-beta
Gene Symbol IFNB1
Biological Activity Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is typically 5-30 pg/mL. The specific activity of Recombinant Human IFN-beta is approximately 2.8 x 108 IU/mg, which is calibrated against human IFN-beta WHO International Standard (NIBSC code: 00/572).Standard (NIBSC code: 00/572).
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Met22-Asn187
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.