Learn More
R&D Systems™ Recombinant Human IFN-gamma Biotinylated Protein
Description
Specifications
Specifications
| Accession Number | CAA31639.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween 80. |
| Molecular Weight (g/mol) | M.W. (Observed): 14-20 kDa, under reducing conditions., M.W. (Theoretical): 16.9 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human IFN-gamma protein Gln24-Gln166, with a N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Conjugate | Biotin |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.