Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IFN-gamma (HEK293-expressed) Protein

Catalog No. 10067IF025
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-067-IF025 25 μg
10-067-IF100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-067-IF025 Supplier R&D Systems™ Supplier No. 10067IF025
This item is not returnable. View return policy

Noncovalent homodimer

The Recombinant Human IFN-gamma (HEK293-expressed) Protein is derived from HEK293. The Recombinant Human IFN-gamma (HEK293-expressed) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_000610.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 3458
Molecular Weight (g/mol) 16.8 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 25 μg
Source Human embryonic kidney cell, HEK293-derived human IFN-gamma protein Met1-Gln166
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.