Learn More
R&D Systems™ Recombinant Human IGF-I R/IGF1R (His-tagged) Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl, EDTA and Tween™-20 with Trehalose. |
| Gene ID (Entrez) | 3480 |
| Molecular Weight (g/mol) | 104 kDa (single chain), 81 kDa ( alpha subunit) and 23 kDa (beta subunit) |
| Name | IGF-I R (Histidine-tagged) |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human IGF-I R/IGF1R protein Met1-Asn932 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Gene Alias | CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.