Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IGFBP-3 Protein

Catalog No. 8874B3025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8874B3025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8874B3025 Supplier R&D Systems™ Supplier No. 8874B3025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IGFBP-3 Protein is derived from Sf 9 (baculovirus). The Recombinant Human IGFBP-3 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P17936
For Use With (Application) Bioactivity
Gene ID (Entrez) 3486
Molecular Weight (g/mol) Observed M.W.: 34-41kDa, reducing conditions, Theoretical M.W.: 29kDa
Quantity 25 μg
Source Spodoptera frugiperda,Sf 9 (baculovirus)-derived human IGFBP-3 protein Gly28-Lys291 Accession &Num; P17936
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Common Name IGFBP-3
Gene Symbol IGFBP3
Biological Activity Measured by its ability to inhibit the biological activity of IGF-I or IGF-II on MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is typically 0.03-0.15μg/mL in the presence of 14ng/mL Recombinant Human IGF-II.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 9 (baculovirus)-derived Gly28-Lys291 Accession # P17936
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.