Learn More
R&D Systems™ Recombinant Human IGSF4C/SynCAM4 Fc Chimera Protein
Description
- Derived from HEK293
- Validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q8NFZ8 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 199731 |
| Molecular Weight (g/mol) | 74 to 97 kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IGSF4C/SynCAM4 protein. Human IGSF4C (Gln25-Tyr323) Accession # Q8NFZ8IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus |
| Storage Requirements | 12 months from date of receipt, -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution 3 months, -20 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.