Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-1 beta/IL-1F2 Protein
SDP

Catalog No. 201LB010CF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-1LB0-10CF 10 μg
20-1LB1-00CF 100 μg
20-1LB0-1MCF 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 20-1LB0-10CF Supplier R&D Systems™ Supplier No. 201LB010/CF
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. Symons, J.A. et al. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 272. The ED50 for this effect is <12 pg/mL. The specific activity of recombinant human IL-1 beta is approximately 1.4 x 105 IU/μg, which is calibrated against recombinant human IL-1 beta WHO International Standard (NIBSC code: 86/680). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Accession Number NP_000567
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. 1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS. See Certificate of Analysis for details.
Gene ID (Entrez) 3553
Molecular Weight (g/mol) 17 kDa
Quantity 10 μg
Source <EM>E. coli</EM>-derived human IL-1 beta/IL-1F2 protein Ala117-Ser269
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. The 1 mg pack size (01M) can be stored for 1 month at 2 to 8 °C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbol IL1B
Conjugate Unconjugated
Cross Reactivity IL-1 beta/IL-1F2
Species Human
Reconstitution Reconstitute at 50-200 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.