Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-1 RI Fc Chimera Protein

Catalog No. 11085RI100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11085RI100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11085RI100 Supplier R&D Systems™ Supplier No. 11085RI100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. When Recombinant Human IL-1 RI Fc Chimera (Catalog # 11085-RI) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Human IL-1 beta/IL-1F2 (Catalog # 201-LB ) binds with an ED50 of 0.600-5.40 ng/mL.

Specifications

Accession Number P14778.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3554
Molecular Weight (g/mol) M.W.-observed: 80-90 kDa, under reducing conditions., M.W.-theroretical: 63 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human IL-1 RI protein Human IL-1 RI (Asp21-Lys336) (C-terminus) Accession # P14778.1 IEGRMD Human IgG1 Fc (Pro100-Lys330) (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1 RI, IL1RT1, IL-1 RT1, IL-1 RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-1 RI
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.