Learn More
Proteintech Group Inc Recombinant Human IL-10 100ug
Supplier: Proteintech Group Inc HZ1145100UG
Animal-free Recombinant Human IL-10 is expressed in human 293 cells as a non-covalent homodimeric glycoprotein with an apparent molecular mass of 17 kDa. This cytokine is produced in a serum-free, chemically defined media. IL-10 is produced by a variety of mammalian cell types which includes T cells, B cells, macrophages, and keratinocytes monocytes. It is an anti-inflammatory cytokine with multiple, pleiotropic effects in inflammation and immunoregulation. Glycosylation contributes to stability in cell growth media and other applications. The cytokine is greater than 95% pure.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.