Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-10, Animal Free Protein

Catalog No. AFL106401M
Change view
Click to view available options
Quantity:
1 mg
100 μg
250 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFL106401M 1 mg
AFL1064050 50 μg
AFL1064100 100 μg
AFL1064250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AFL106401M Supplier R&D Systems™ Supplier No. AFL106401M
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is 0.0750-0.750 ng/mL.

The specific activity of recombinant human IL-10 is >2.50 x 106 units/mg, which is calibrated against the human IL-10 WHO Reference Reagent (NIBSC code: 93/722).

Specifications

Accession Number P22301.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3586
Molecular Weight (g/mol) M.W.-observed: 18 kDa, reducing conditions, M.W.-theroretical: 19 kDa
Quantity 1 mg
Source E. coli-derived human IL-10 protein Ser19-Asn178, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.