Learn More
R&D Systems™ Recombinant Human IL-10, Animal Free Protein
Description
The specific activity of recombinant human IL-10 is >2.50 x 106 units/mg, which is calibrated against the human IL-10 WHO Reference Reagent (NIBSC code: 93/722).
Specifications
Specifications
| Accession Number | P22301.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 3586 |
| Molecular Weight (g/mol) | M.W.-observed: 18 kDa, reducing conditions, M.W.-theroretical: 19 kDa |
| Quantity | 100 μg |
| Source | E. coli-derived human IL-10 protein Ser19-Asn178, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.