Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-10 Protein

Catalog No. 1064ILB050
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1064ILB050 50 μg
1064ILB010 10 μg
1064ILB01M 1 mg
1064ILB250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 1064ILB050 Supplier R&D Systems™ Supplier No. 1064ILB050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is 0.075-0.750 ng/mL.

Specifications

Accession Number P22301.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3586
Molecular Weight (g/mol) M.W.-observed: 18 kDa, under reducing conditions., M.W.-theroretical: 19 kDa
Quantity 50 μg
Source E. coli-derived human IL-10 protein Ser19-Asn178, with a N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.