Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-10 Protein

Catalog No. 217ILB005
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-7IL-B005 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-7IL-B005 Supplier R&D Systems™ Supplier No. 217ILB005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-10 Protein is derived from Sf 21 (stably transfected). The Recombinant Human IL-10 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P22301
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Gene ID (Entrez) 3586
Molecular Weight (g/mol) 18.6kDa
Name IL-10
Preparation Method Reconstitute at 50μg/mL in PBS containing at least 0.1% human or bovine serum albumin.
Quality Control Testing SDS-PAGE: 18kDa, reducing conditions
Quantity 5 μg
Source Spodoptera frugiperda,Sf 21 (stably transfected)-derived human IL-10 protein Met1-Asn178
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ser19
Structural Form Noncovalently-linked homodimer
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.