Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-11 Protein
SDP

Catalog No. 218IL005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS and EDTA.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
218IL005 5 μg Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
218IL005CF 5 μg Supplied as a 0.2μm filtered solution in PBS and EDTA.
218IL025 25 μg Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
218IL025CF 25 μg Supplied as a 0.2μm filtered solution in PBS and EDTA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 218IL005 Supplier R&D Systems™ Supplier No. 218IL005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using T11 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.02-0.12 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 3589
Molecular Weight (g/mol) 19 kDa
Name IL-11
Quantity 5 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-11 protein Pro22-Leu199
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.