Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-12 (linked heterodimer) Protein

Catalog No. 10018IL100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-018-IL100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-018-IL100 Supplier R&D Systems™ Supplier No. 10018IL100
Add to Cart
Add to Cart
This item is not returnable. View return policy

GS Linker, Defined heterodimer stoichiometry.

  • Measured in a cell proliferation assay using PHA-stimulated human T lymphoblasts
  • ED50 for this effect is 0.01 to 0.05 ng/mL

Specifications

Accession Number P29460.1 (IL-12p40), P29459.2 (IL-12p35)
For Use With (Application) Bioactivity
Gene ID (Entrez) 3592
Molecular Weight (g/mol) Observed M.W.: 63-75 kDa, under reducing conditions.
Name CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human IL-12 protein Human IL-12p40 (Ile23-Ser328) (N-terminus) Accession # P29460.1 GSGSSRGGSGSGGSGGGGSK Human IL-12p35 (Arg23-Ser219) (N-terminus) Accession # P29459.2(C-terminus)
Storage Requirements 12 months from date of receipt, ≤-20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, ≤-20 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100-200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.