Learn More
R&D Systems™ Recombinant Human IL-12 R beta 1 Fc Chimera Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3594 |
| Molecular Weight (g/mol) | 84.5 kDa (monomer) |
| Name | IL-12 R beta 1 Fc Chimera |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line,NS0-derived human IL-12 R beta 1 protein, (N-terminus) Human IL-12 R beta 1 (Cys24-Glu540) Accession &Num; P42701 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.