Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-12 R beta 1 His Protein
SDP

Catalog No. NBP25208505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25208505 0.05 mg
NBP25208525 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25208505 Supplier Novus Biologicals™ Supplier No. NBP2520850.05MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 24-545 of Human IL-12 R beta 1 The Recombinant Human IL-12 R beta 1 Protein is derived from Baculovirus. The Recombinant Human IL-12 R beta 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH 7.4) containing 20% glycerol, 1mM EDTA, 0.1mM PMSF.
Gene ID (Entrez) 3594
Molecular Weight (g/mol) TMW: 58.4kDa
Quantity 0.05 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL-12R subunit beta-1, IL12RB, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Common Name IL-12 R beta 1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.