Learn More
R&D Systems™ Recombinant Human IL-13 R alpha2 Fc Alexa Fluor™ 647 Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | NP_000631.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 3598 |
| Molecular Weight (g/mol) | M.W.-observed: 75-95 kDa, under reducing conditions., M.W.-theroretical: 64 kDa (monomer) |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human IL-13 R alpha 2 protein Human IL-13 R alpha 2 (Cys22-Leu342) (C-terminus) Accession # NP_000631.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13 R, IL13R alpha 2, IL-13 R subunit alpha-2, IL13RA2, IL-13 RA2, IL-13 R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein |
| Conjugate | Alexa Fluor 647 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.