Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-15 Protein
SDP

Catalog No. 247ILB005CF Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
25 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
247ILB005CF 5 μg
247ILB025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 247ILB005CF Supplier R&D Systems™ Supplier No. 247ILB005/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 0.3-2.6 ng/mL. The specific activity of Recombinant Human IL-15 is approximately 4.5 x 105 U/μg, which is calibrated against recombinant human IL-15 WHO International Standard (NIBSC code: 95/554). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Accession Number P40933.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS. Reconstitute at 100μg/mL in sterile PBS.
Gene ID (Entrez) 3600
Molecular Weight (g/mol) 10 kDa, under reducing conditions.
Quantity 5 μg
Source E. coli-derived human IL-15 protein Asn49-Ser162
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 °C to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias IL15, IL-15MGC9721, interleukin 15, interleukin-15
Common Name IL-15
Gene Symbol IL15
Biological Activity Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is typically 0.3-2.6ng/mL. The specific activity of Recombinant Human IL-15 is approximately 4.5 x 105 U/μg, which is calibrated against recombinant human IL-15 WHO International Standard (NIBSC code: 95/554).
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Asn49-Ser162
Product Line R&D Systems
Purity or Quality Grade >97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.