Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-16 Protein

Catalog No. 316IL002CF
Change view
Click to view available options
Quantity:
2 μg
25 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in PBS.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
316IL002CF 2 μg Supplied as a 0.2 μm filtered solution in PBS.
316IL002 2 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
316IL025 25 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
316IL025CF 25 μg Supplied as a 0.2 μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 316IL002CF Supplier R&D Systems™ Supplier No. 316IL002/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-16 Protein is derived from E. coli. The Recombinant Human IL-16 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3603
Molecular Weight (g/mol) 13.3 kDa
Name IL-16
Quantity 2 μg
Source E. coli-derived human IL-16 protein Pro2-Ser130
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.