Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-17A Biotinylated Protein
SDP

Catalog No. BT7955B025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT7955B025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT7955B025 Supplier R&D Systems™ Supplier No. BT7955B025
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Biotinylated

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human IL-17/IL-17A (Catalog # BT7955B) binds to Recombinant Human IL-17RA/IL-17R Fc Chimera Protein (Catalog # 177-IR ) with a ED50 of 2.00-20.0 ng/mL.

Specifications

Accession Number Q16552.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HCl with Trehalose.
Gene ID (Entrez) 3605
Molecular Weight (g/mol) MolecularWeight-observed: 12-25 kDa, under reducing conditions., MolecularWeight-theroretical: 15.1 kDa
Quantity 25 μg
Source Human embryonic kidney cell, HEK293-derived human IL-17/IL-17 A protein Gly24-Ala155
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17 A, IL-17 Acytotoxic T-lymphocyte-associated protein 8, IL-17 CTLA-8, IL17interleukin-17 A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17 A
Conjugate Biotin
Reconstitution Reconstitute at 100 μg/mL in HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-17/IL-17 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.