Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-17A/F Heterodimer Biotinylated Protein

Catalog No. BT5837025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT5837025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT5837025 Supplier R&D Systems™ Supplier No. BT5837025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-17A/F Heterodimer Biotinylated Protein is derived from CHO. The Recombinant Human IL-17A/F Heterodimer Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 3605
Molecular Weight (g/mol) Theoretical M.W.: 15kDa (unlabeled IL-17A), 15kDa (unlabeled IL-17F)
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human IL-17A/F Heterodimer protein, (N-terminus) Human IL-17A (Gly24-Ala155) Accession &Num; Q16552 Human IL-17F (Arg21-GLN163) Accession &Num; Q96PD4 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias CTLA8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, Cytotoxic T-Lymphocyte-Associated Protein 8, IL17, IL-17, IL-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A, interleukin-17A
Common Name IL-17A/F Heterodimer
Biological Activity Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. The ED50 for this effect is typically 0.4-2ng/mL.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Human IL-17A (Gly24-Ala155) Accession # Q16552, N-terminus; Accession # Q96PD4, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.