Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-17B (CHO) Protein
SDP

Catalog No. 8129IL010CF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-29I-L010CF 10 μg
81-29I-L100CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 81-29I-L010CF Supplier R&D Systems™ Supplier No. 8129IL010/CF
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to induce IL-8 secretion by HepG2 human hepatocellular carcinoma cells. ED50 for this effect is 0.4 to 2 μg/mL.

Specifications

Accession Number Q9UHF5
For Use With (Application) Bioactivity
Gene ID (Entrez) 27190
Molecular Weight (g/mol) 18 kDa (monomer)
Name Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-17B protein Gln21-Phe180
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol IL17B
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in Sodium Phosphate and NaCl.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.