Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-17F (NS0-expressed) Protein

Catalog No. 1335INS100C
Change view
Click to view available options
Quantity:
10 μg
100 μg
25 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-35I-NS100C 100 μg
1335INS025C 25 μg
13-35I-NS010C 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 13-35I-NS100C Supplier R&D Systems™ Supplier No. 1335INS100/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-17F (NS0-expressed) Protein is derived from NS0. The Recombinant Human IL-17F (NS0-expressed) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q96PD4
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 um filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 112744
Molecular Weight (g/mol) 15.7 kDa (monomer)
Name IL-17F
Quantity 100 μg
Source Mouse myeloma cell line,NS0-derived human IL-17F protein Arg31-Gln163,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Symbol IL17F
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 ug/mL in sterile 4 mM HCl.
Buffer Lyophilized from a 0.2 um filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.10 EU / 1 ug of the protein by the LAL method.
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.