Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-17RA/IL-17R His-tag Protein

Catalog No. 11234IR100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11234IR100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11234IR100 Supplier R&D Systems™ Supplier No. 11234IR100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its ability to inhibit IL-17-induced IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells. The ED50 for this effect is 0.050-1.00 μg/mL in the presence of 10.0 ng/mL recombinant human IL-17 (Catalog # 7955-IL ).

Specifications

Accession Number NP_055154.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 23765
Molecular Weight (g/mol) M.W.-observed: 60-68 kDa, under reducing conditions., M.W.-theroretical: 34 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-17 RA/IL-17 R protein Leu33-Trp320, with a C-terminal 6-His tag.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17 R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17 RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-17 RA/IL-17 R
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.