Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-18/IL-1F4 Protein

Catalog No. 9124IL500CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-24I-L500CF 500 μg
91-24I-L010CF 10 μg
91-24I-L050CF 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 91-24I-L500CF Supplier R&D Systems™ Supplier No. 9124IL500/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-18/IL-1F4 Protein is derived from E. coli. The Recombinant Human IL-18/IL-1F4 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q14116
For Use With (Application) Bioactivity
Gene ID (Entrez) 3606
Molecular Weight (g/mol) Observed M.W.: 19kDa, reducing conditions, Theoretical M.W.: 18kDa
Quantity 500 μg
Source E. coli-derived human IL-18/IL-1F4 protein Tyr37-Asp193
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Common Name IL-18/IL-1F4
Gene Symbol IL18
Biological Activity Measured by its ability to induce IFN-gamma secretion by KG-1 human acute myelogenous leukemia cells in the presence of TNF-alpha . The ED50 for this effect is typically 1.5-9ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Tyr37-Asp193
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.