Learn More
R&D Systems™ Recombinant Human IL-18/IL-1F4 Protein
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2μm filtered solution in phosphate-buffered saline (PBS) with stabilizers and preservatives. |
| Molecular Weight (g/mol) | Predicted MW: Observed MW: PredictedMoleMass: 18kDa |
| Quantity | 25 μg |
| Source | E. coli-derived human IL-18/IL-1F4 protein 37-193 aa |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Unconjugated |
| Reconstitution | Sterile PBS to a final concentration of 0.5mg/mL. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.