Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-18 R beta/IL-1 R7 Fc Chimera

Catalog No. 118AP100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
118AP100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 118AP100 Supplier R&D Systems™ Supplier No. 118AP100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit the IL-18-induced response of KG-1 human acute myelogenous leukemia cells in the presence of soluble Recombinant Human IL-18 R alpha/IL-1 R5 Fc Chimera (Catalog # 816-LR ). The ED50 for this effect is 2-6 μg/mL in the presence of 30 μg/mL of soluble IL-18 R alpha/Fc Chimera and 40 ng/mL of human IL-18.

Specifications

Accession Number O95256
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8807
Molecular Weight (g/mol) M.W. (Observed): 78-93 kDa, reducing conditions; M.W. (theoretical): 65.1 kDa (monomer)
Name IL-18 R beta/IL-1 R7 Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human IL-18 R beta/IL-1 R7 protein Human IL-18 R beta (Phe20 - Arg356) (N-terminus) Accession # O95256 DIEGRMD Human IgG1 (Pro100 - Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol IL18RAP
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-18 R beta/IL-1 R7/ACPL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.