Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-2, Animal-Free Protein

Catalog No. AFL202050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFL202050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFL202050 Supplier R&D Systems™ Supplier No. AFL202050
This item is not returnable. View return policy

Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells

Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295. The ED50 for this effect is 0.05-0.25 ng/mL. The specific activity of Recombinant Human IL-2 is approximately 2.1 x 104 IU/μg, which is calibrated against recombinant human IL-2 WHO International Standard (NIBSC code: 86/500).

Specifications

Accession Number P60568.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) M.W. (theoretical): 15 kDa
Name aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Quantity 50 μg
Source E. coli-derived human IL-2 protein Ala21-Thr153, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias aldesleukin, IL2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Gene Symbol IL2
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 0.2 mg/mL in sterile 100 mM Acetic Acid. Alternatively, reconstitute at 0.1 mg/mL in sterile deionized water and use within 24 hours, store (at 2°C to 8°C.)
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.